siRNA Details
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009259.5 | NP_033285.1 | 2 | 2341 | ||
GenBank mRNA | AK077764.1 | - | 2 | 2332 | ||
AK134098.1 | - | 2 | 2336 | |||
S70677.1 | - | 1 | 2233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
