siRNA Details
Gene Symbol: Swap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009302.3 | NP_033328.3 | 11 | 1742 | ||
GenBank mRNA | AF053974.1 | - | 11 | 1634 | ||
AK019882.1 | - | 11 | 1686 | |||
AK146773.1 | - | 11 | 1734 | |||
AK150621.1 | - | 5 | 714 | |||
AK163357.1 | - | 4 | 552 | |||
AK172990.1 | - | 11 | 1679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
