siRNA Details
Gene Symbol: Syngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009303.2 | NP_033329.1 | 4 | 605 | ||
GenBank mRNA | AJ002306.1 | - | 4 | 585 | ||
AK010442.1 | - | 4 | 572 | |||
AK153096.1 | - | 4 | 572 | |||
AK153187.1 | - | 4 | 572 | |||
AK156134.1 | - | 4 | 589 | |||
AK190600.1 | - | 1 | 66 | |||
AK190974.1 | - | 1 | 67 | |||
AK192355.1 | - | 1 | 67 |

More Information
Other Species Targeted

Gene Ontology Categories:
