siRNA Details
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009323.2 | NP_033349.2 | 8 | 3491 | ||
GenBank mRNA | AF041822.1 | - | 8 | 3394 | ||
AK132578.1 | - | 8 | 3492 | |||
AK137458.1 | - | 8 | 3288 | |||
AY662679.1 | - | 8 | 3412 |

More Information
Other Species Targeted

Gene Ontology Categories:
