siRNA Details
Gene Symbol: HOXA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024014.3 | NP_076919.1 | 2 | 669 | ||
GenBank mRNA | BC069497.1 | - | 2 | 644 | 7262260 | |
BC104915.1 | - | 2 | 669 | 8143918 | ||
BC104917.1 | - | 2 | 669 | 8143920 | ||
BG749730.1 | - | 2 | 413 |
