siRNA Details
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025261.1 | NP_001020432.1 | 2, 3 | 332 | ||
NM_001025262.1 | NP_001020433.1 | 2, 3 | 332 | |||
NM_001025263.1 | NP_001020434.1 | 2, 3 | 235 | |||
NM_001025264.1 | NP_001020435.1 | 2, 3 | 235 | |||
NM_009412.2 | NP_033438.1 | 2, 3 | 235 | |||
GenBank mRNA | AK005456.1 | - | 2, 3 | 216 | ||
AK032111.1 | - | 2, 3 | 198 | |||
AK150935.1 | - | 2, 3 | 198 | |||
AK151471.1 | - | 2, 3 | 196 | |||
AK151744.1 | - | 2, 3 | 196 | |||
AK152543.1 | - | 2, 3 | 196 | |||
AK153266.1 | - | 2, 3 | 196 | |||
AK153292.1 | - | 2, 3 | 206 | |||
AK161753.1 | - | 2, 3 | 197 | |||
AK168379.1 | - | 2, 3 | 191 | |||
AK168817.1 | - | 2, 3 | 199 | |||
AK169751.1 | - | 2, 3 | 228 | |||
AK170725.1 | - | 2, 3 | 181 | |||
AK171050.1 | - | 2, 3 | 176 | |||
AY048852.1 | - | 2, 3 | 324 | |||
BC002036.1 | - | 2, 3 | 215 | 3593963 | ||
BC004068.1 | - | 2, 3 | 168 | 3590969 | ||
BC094018.1 | - | 2, 3 | 191 | 3674995 | ||
BE634665.1 | - | 2, 3 | 217 | |||
BY282453.1 | - | 2, 3 | 235 | |||
CA480795.1 | - | 2, 3 | 369 | |||
U44426.1 | - | 2, 3 | 164 |

More Information
Other Species Targeted

Gene Ontology Categories:
