siRNA Details
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009348.3 | NP_033374.2 | 10 | 1774 | ||
GenBank mRNA | AK010948.1 | - | 4 | 1338 | ||
BC138888.1 | - | 10 | 1701 | 8861910 | ||
BC138889.1 | - | 10 | 1702 | 8861911 | ||
X99806.2 | - | 10 | 1775 |
