siRNA Details
Gene Symbol: GPER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039966.1 | NP_001035055.1 | 3 | 1899 | ||
NM_001098201.1 | NP_001091671.1 | 2 | 1572 | |||
NM_001505.2 | NP_001496.1 | 2 | 1775 | |||
GenBank mRNA | AB451427.1 | - | 1 | 1015 | ||
AF015257.1 | - | 2 | 1706 | |||
AK289588.1 | - | 2 | 1546 | |||
BC011634.1 | - | 1 | 1106 | 4079700 | ||
CR541655.1 | - | 1 | 1015 | |||
CR541741.1 | - | 1 | 1015 | |||
U58828.1 | - | 2 | 964 | |||
U63917.1 | - | 2 | 1519 | |||
X98510.1 | - | 1 | 1188 | |||
Y08162.1 | - | 1 | 1015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
