siRNA Details
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005233.5 | NP_005224.2 | 3 | 440 | ||
NM_182644.2 | NP_872585.1 | 3 | 440 | |||
XM_005264715.2 | XP_005264772.1 | 3 | 365 | |||
XM_005264716.2 | XP_005264773.1 | 3 | 435 | |||
GenBank mRNA | AF213459.1 | - | 3 | 440 | ||
AF213460.1 | - | 3 | 302 | |||
AK291411.1 | - | 3 | 273 | |||
BC063282.1 | - | 3 | 344 | 30347891 | ||
M83941.1 | - | 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
