siRNA Details
Gene Symbol: Tsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009440.2 | NP_033466.1 | 1 | 163 | ||
GenBank mRNA | BC048493.1 | - | 1 | 163 | 6705048 | |
HQ433540.1 | - | 2 | 274 | |||
HQ433541.1 | - | 2 | 378 | |||
HQ433542.1 | - | 2 | 273 | |||
HQ433544.1 | - | 1 | 463 | |||
HQ433547.1 | - | 1 | 363 | |||
X99796.1 | - | 1 | 148 |
