siRNA Details
Gene Symbol: Tgoln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009444.1 | NP_033470.1 | 4 | 1786 | ||
GenBank mRNA | D50032.1 | - | 4 | 1786 |
Gene Symbol: Tgoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009443.3 | NP_033469.1 | 4 | 2009 | ||
GenBank mRNA | AK041302.1 | - | 4 | 1759 | ||
AK076586.1 | - | 4 | 2010 | |||
AK133559.1 | - | 4 | 1756 | |||
AK162574.1 | - | 4 | 1759 | |||
AK218314.1 | - | 1 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
