siRNA Details
Gene Symbol: Tubb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009451.3 | NP_033477.2 | 2 | 414 | ||
GenBank mRNA | AK013717.1 | - | 2 | 232 | ||
BC049112.1 | - | 2 | 414 | 6415577 | ||
BC054831.1 | - | 2 | 213 | 6306945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
