siRNA Details
Gene Symbol: Ugt2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009467.3 | NP_033493.3 | 2 | 770 | ||
GenBank mRNA | AK136480.1 | - | 2 | 771 | ||
BC028262.1 | - | 2 | 761 | 5102040 | ||
X06358.1 | - | 2 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
