siRNA Details
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005233.5 | NP_005224.2 | 6, 7 | 1667 | ||
NM_182644.2 | NP_872585.1 | 6, 7 | 1667 | |||
XM_005264716.2 | XP_005264773.1 | 6, 7 | 1662 | |||
GenBank mRNA | AF213459.1 | - | 6, 7 | 1667 | ||
AF213460.1 | - | 6, 7 | 1529 | |||
AK291411.1 | - | 6, 7 | 1500 | |||
BC063282.1 | - | 6, 7 | 1571 | 30347891 | ||
M83941.1 | - | 6, 7 | 1542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
