siRNA Details
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009448.4 | NP_033474.1 | 4 | 1880 | ||
GenBank mRNA | AK013029.1 | - | 2 | 1036 | ||
AK145567.1 | - | 4 | 1480 | |||
AK167654.1 | - | 4 | 1450 | |||
AK167759.1 | - | 4 | 1413 | |||
AK191962.1 | - | 1 | 258 | |||
AK202573.1 | - | 1 | 277 | |||
M13441.1 | - | 4 | 1392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
