siRNA Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145312.1 | NP_001138784.1 | 3 | 533 | ||
NM_005240.1 | NP_005231.1 | 3 | 331 | |||
XM_006711210.2 | XP_006711273.1 | 3 | 514 | |||
GenBank mRNA | AF218540.1 | - | 2 | 244 | ||
AK304789.1 | - | 3 | 337 | |||
AK316274.1 | - | 3 | 533 | |||
BC022868.1 | - | 3 | 331 | |||
L16464.1 | - | 3 | 310 |
Gene Symbol: ETV3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004341.2 | NP_001004341.1 | 2 | 541 | ||
GenBank mRNA | AK131392.1 | - | 2 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
