siRNA Details
Gene Symbol: Atrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009530.2 | NP_033556.2 | 4 | 484 | ||
GenBank mRNA | AF026032.1 | - | 4 | 451 | ||
AK032955.1 | - | 1 | 13 | |||
AK088095.1 | - | 6 | 879 | |||
AK135019.1 | - | 5 | 755 | |||
AK172134.1 | - | 4 | 302 |
