siRNA Details
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009453.3 | NP_033479.2 | 8 | 969 | ||
GenBank mRNA | CN700359.1 | - | 1 | 190 | ||
D45205.1 | - | 8 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
