siRNA Details
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009507.3 | NP_033533.1 | 3 | 953 | ||
GenBank mRNA | AK159238.1 | - | 3 | 974 | ||
AK171264.1 | - | 3 | 955 | |||
AK171604.1 | - | 3 | 963 | |||
U12570.1 | - | 3 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
