siRNA Details
Gene Symbol: Zbtb7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009565.4 | NP_033591.2 | 2 | 1018 | ||
GenBank mRNA | AK149976.1 | - | 2 | 966 | ||
AK155784.1 | - | 2 | 978 | |||
AK171443.1 | - | 3 | 1663 | |||
BC046533.1 | - | 1 | 857 | 4946243 | ||
BC089185.1 | - | 1 | 652 | |||
BC091651.1 | - | 2 | 1018 | 6309645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
