siRNA Details
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036833.1 | - | 6 | 920 | ||
AK040268.1 | - | 5 | 653 | |||
AK077815.1 | - | 6 | 1217 | |||
AK154175.1 | - | 6 | 942 | |||
AK220235.1 | - | 6 | 886 | |||
AY536269.1 | - | 3 | 230 | |||
BC138789.1 | - | 5 | 451 | 8861811 | ||
L03547.1 | - | 5 | 436 | |||
S74708.1 | - | 3 | 497 |
