siRNA Details
Gene Symbol: Dnajc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009584.4 | NP_033610.1 | 14 | 1627 | ||
GenBank mRNA | AK131964.1 | - | 14 | 1648 | ||
AK167991.1 | - | 14 | 1648 | |||
BC021755.1 | - | 13 | 3372 | |||
BC052027.1 | - | 14 | 1654 | 5718637 | ||
D63784.1 | - | 14 | 1626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
