siRNA Details
Gene Symbol: Adnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009628.2 | NP_033758.2 | 4 | 1747 | ||
GenBank mRNA | AF068198.1 | - | 1 | 413 | ||
AK129214.1 | - | 4 | 1798 | |||
AK134786.1 | - | 3 | 1269 | |||
BC050833.1 | - | 1 | 696 | |||
BC057666.1 | - | 1 | 805 | |||
BC090840.1 | - | 1 | 943 |

More Information
Other Species Targeted

Gene Ontology Categories:
