siRNA Details
Gene Symbol: ERBB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042599.1 | NP_001036064.1 | 12 | 1472 | ||
NM_005235.2 | NP_005226.1 | 12 | 1472 | |||
XM_005246376.2 | XP_005246433.1 | 12 | 1991 | |||
XM_005246377.2 | XP_005246434.1 | 12 | 1991 | |||
XM_006712364.2 | XP_006712427.1 | 12 | 1991 | |||
XM_017003577.1 | XP_016859066.1 | 13 | 2069 | |||
XM_017003578.1 | XP_016859067.1 | 13 | 2069 | |||
XM_017003579.1 | XP_016859068.1 | 13 | 2069 | |||
XM_017003580.1 | XP_016859069.1 | 13 | 2069 | |||
XM_017003581.1 | XP_016859070.1 | 13 | 2069 | |||
XM_017003582.1 | XP_016859071.1 | 8 | 1104 | |||
GenBank mRNA | AB209697.1 | - | 2 | 176 | ||
AK308606.1 | - | 12 | 1428 | |||
BC112199.1 | - | 12 | 1413 | 8327667 | ||
BC143741.1 | - | 12 | 1401 | 9052255 | ||
BC143747.1 | - | 12 | 1401 | 9052261 | ||
BC143749.1 | - | 12 | 1401 | 9052263 | ||
L07868.1 | - | 12 | 1407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
