siRNA Details
Gene Symbol: Zfp185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109043.1 | NP_001102513.1 | 13 | 1088 | ||
NM_009549.3 | NP_033575.3 | 13 | 1091 | |||
GenBank mRNA | AK014497.1 | - | 2 | 139 | ||
AK048188.1 | - | 13 | 1069 | |||
BC039772.1 | - | 10 | 676 | 4191053 | ||
EF494744.1 | - | 13 | 1071 | |||
U46687.1 | - | 12 | 917 |
