siRNA Details
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149288.1 | - | 3 | 447 | ||
BC114999.1 | - | 3 | 404 | 6506501 | ||
BC145270.1 | - | 3 | 423 | 9053824 | ||
U69613.1 | - | 3 | 412 | |||
U69614.1 | - | 3 | 496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
