siRNA Details
Gene Symbol: Ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177602.1 | NP_001171073.1 | 3 | 549 | ||
NM_001177604.1 | NP_001171075.1 | 3 | 362 | |||
NM_001177605.1 | NP_001171076.1 | 3 | 340 | |||
NM_009647.5 | NP_033777.1 | 2 | 307 | |||
GenBank mRNA | AB020239.1 | - | 2 | 271 | ||
AK134600.1 | - | 3 | 552 | |||
AK144439.1 | - | 2 | 277 | |||
AK154374.1 | - | 3 | 366 | |||
AK166975.1 | - | 2 | 229 | |||
BC086663.1 | - | 2 | 255 | 30649610 | ||
BC086791.1 | - | 2 | 271 | |||
BY718404.1 | - | 3 | 550 | |||
BY740484.1 | - | 3 | 341 | |||
CJ087385.1 | - | 2 | 307 | |||
D85036.1 | - | 2 | 271 |
