siRNA Details
Gene Symbol: Zfp185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109043.1 | NP_001102513.1 | 19 | 1811 | ||
NM_009549.3 | NP_033575.3 | 19 | 1814 | |||
GenBank mRNA | AK048188.1 | - | 19 | 1792 | ||
BC039772.1 | - | 16 | 1399 | 4191053 | ||
EF494744.1 | - | 19 | 1794 | |||
U46687.1 | - | 18 | 1640 |
