siRNA Details
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009604.3 | NP_033734.3 | 7 | 712 | ||
GenBank mRNA | AK047974.1 | - | 7 | 713 | ||
X03818.1 | - | 7 | 709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
