siRNA Details
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301230.1 | NP_001288159.1 | 7 | 2684 | ||
NM_001301232.1 | NP_001288161.1 | 6 | 2577 | |||
NM_023635.6 | NP_076124.1 | 6 | 2535 | |||
GenBank mRNA | AK008798.1 | - | 1 | 707 | ||
AK030529.1 | - | 6 | 2481 |
Gene Symbol: Bcl2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289716.1 | NP_001276645.1 | 2 | 728 | ||
NM_001289717.1 | NP_001276646.1 | 2 | 573 | |||
NM_001289739.1 | NP_001276668.1 | 2 | 728 | |||
NM_009743.5 | NP_033873.3 | 2 | 522 | |||
GenBank mRNA | AK146461.1 | - | 2 | 678 | ||
AK150028.1 | - | 2 | 465 | |||
AK172250.1 | - | 2 | 489 | |||
BC089016.1 | - | 2 | 476 | 30606568 | ||
BC089017.1 | - | 1 | 313 | 30605282 | ||
L35049.1 | - | 2 | 526 | |||
U10100.1 | - | 1 | 284 | |||
U10101.1 | - | 1 | 284 | |||
U10102.1 | - | 2 | 521 | |||
U51277.1 | - | 2 | 661 | |||
U51278.1 | - | 1 | 386 | |||
U51279.1 | - | 2 | 519 | |||
X83574.1 | - | 1 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
