siRNA Details
Gene Symbol: Anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009672.3 | NP_033802.2 | 2 | 272 | ||
GenBank mRNA | AF022957.1 | - | 2 | 181 | ||
AK020753.1 | - | 2 | 234 | |||
BC062899.1 | - | 2 | 187 | 6415168 | ||
CJ150714.1 | - | 2 | 272 | |||
U73478.1 | - | 2 | 139 |
Gene Symbol: Anp32-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY702092.1 | - | 1 | 114 |

More Information
Other Species Targeted

Gene Ontology Categories:
