siRNA Details
Gene Symbol: Aox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009676.2 | NP_033806.2 | 33 | 3657 | ||
GenBank mRNA | AB017482.1 | - | 33 | 3723 | ||
AF076216.1 | - | 33 | 3622 | |||
BC026132.1 | - | 33 | 3649 | 4011903 |
Gene Symbol: Aox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008419.2 | NP_001008419.1 | 33 | 3735 | ||
GenBank mRNA | AY187018.2 | - | 33 | 3735 | ||
AY665589.1 | - | 33 | 3642 | |||
BC150826.1 | - | 33 | 3662 | 9087737 |

More Information
Other Species Targeted

Gene Ontology Categories:
