siRNA Details
Gene Symbol: Car2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009801.4 | NP_033931.4 | 7 | 1517 | ||
GenBank mRNA | AK002333.1 | - | 7 | 1252 | ||
AK002498.1 | - | 6 | 985 | |||
AK146015.1 | - | 7 | 1254 | |||
BC044806.1 | - | 7 | 1238 | |||
BC055291.1 | - | 7 | 1237 | 6479187 | ||
M25944.1 | - | 1 | 470 | |||
U37706.1 | - | 1 | 413 |

More Information
Other Species Targeted

Gene Ontology Categories:
