siRNA Details
Gene Symbol: Ccne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037134.2 | NP_001032211.1 | 9 | 1008 | ||
NM_001282943.1 | NP_001269872.1 | 9 | 921 | |||
NM_009830.3 | NP_033960.1 | 9 | 918 | |||
GenBank mRNA | AF091432.1 | - | 8 | 813 | ||
AF106691.1 | - | 7 | 788 | |||
AK010929.1 | - | 9 | 848 | |||
AK011883.1 | - | 4 | 480 | |||
AK031116.1 | - | 9 | 845 | |||
AK037791.1 | - | 9 | 848 | |||
AK158720.1 | - | 9 | 1434 | |||
AK167428.1 | - | 9 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
