siRNA Details
Gene Symbol: OR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012369.2 | NP_036501.2 | 1 | 930 | ||
XM_011516019.2 | XP_011514321.1 | 4 | 1476 | |||
XM_011516020.2 | XP_011514322.1 | 4 | 1251 | |||
GenBank mRNA | BC104975.1 | - | 1 | 930 | 8143978 |
Gene Symbol: OR2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004685.1 | NP_001004685.1 | 1 | 886 | ||
GenBank mRNA | BC136812.1 | - | 1 | 953 | 9020802 | |
BC136813.1 | - | 1 | 953 | 9020803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
