siRNA Details
Gene Symbol: Cplx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009946.3 | NP_034076.1 | 4 | 548 | ||
GenBank mRNA | CX241609.1 | - | 4 | 548 | ||
D38613.1 | - | 3 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
