siRNA Details
Gene Symbol: ZSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017894.5 | NP_060364.4 | 3 | 672 | ||
XM_017022392.1 | XP_016877881.1 | 4 | 797 | |||
GenBank mRNA | AK000602.1 | - | 4 | 645 | ||
BC041620.1 | - | 4 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
