siRNA Details
Gene Symbol: Cyp2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009997.2 | NP_034127.2 | 2, 3 | 371 | ||
GenBank mRNA | BC010761.1 | - | 2, 3 | 362 | 4239813 | |
BC063778.1 | - | 2, 3 | 394 | 5044482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
