siRNA Details
Gene Symbol: Cyp2b9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010000.2 | NP_034130.1 | 9 | 1527 | ||
GenBank mRNA | BC120525.1 | - | 9 | 1524 | 40129448 | |
BC120527.1 | - | 9 | 1525 | 40129450 | ||
M21855.1 | - | 9 | 1503 |
