siRNA Details
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 24 | 5495 | ||
NM_001291234.1 | NP_001278163.1 | 23 | 5134 | |||
NM_009986.4 | NP_034116.3 | 24 | 4900 | |||
GenBank mRNA | AK220173.1 | - | 23 | 4890 | ||
BC090847.1 | - | 24 | 4672 | 30547458 | ||
X75013.1 | - | 20 | 4690 |

More Information
Other Species Targeted

Gene Ontology Categories:
