siRNA Details
Gene Symbol: Cx3cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009987.4 | NP_034117.3 | 2 | 1111 | ||
GenBank mRNA | AF102269.1 | - | 2 | 1105 | ||
AK035902.1 | - | 2 | 1086 | |||
AK044557.1 | - | 2 | 1112 | |||
AK045634.1 | - | 2 | 1099 | |||
AK137388.1 | - | 2 | 1099 | |||
AK140010.1 | - | 2 | 1085 | |||
BC012653.1 | - | 2 | 1067 | 4017224 |
