siRNA Details
Gene Symbol: Cst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146333.1 | - | 3 | 567 | ||
AK166430.1 | - | 3 | 530 | |||
BC002072.1 | - | 3 | 484 | 3593373 | ||
M59470.1 | - | 3 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
