siRNA Details
Gene Symbol: Cux1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291233.1 | NP_001278162.1 | 24 | 5875 | ||
NM_001291234.1 | NP_001278163.1 | 23 | 5514 | |||
NM_009986.4 | NP_034116.3 | 24 | 5280 | |||
GenBank mRNA | AK043431.1 | - | 1 | 224 | ||
X75013.1 | - | 20 | 5071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
