siRNA Details
Gene Symbol: Cyp2c37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010001.2 | NP_034131.2 | 7, 8 | 1173 | ||
GenBank mRNA | AF047542.1 | - | 1157 | |||
AK005017.1 | - | 7, 8 | 1174 | |||
BC057912.1 | - | 1158 | 5052873 |
Gene Symbol: Cyp2c54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206537.2 | NP_996260.1 | 7, 8 | 1159 | ||
GenBank mRNA | AI047349.1 | - | 4, 5 | 595 | ||
BC043929.1 | - | 6, 7 | 941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
