siRNA Details
Gene Symbol: Sparcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010097.4 | NP_034227.3 | 3 | 796 | ||
GenBank mRNA | AK134608.1 | - | 3 | 437 | ||
AK158725.1 | - | 3 | 303 | |||
BC003759.1 | - | 3 | 441 | 3486162 | ||
U64827.1 | - | 3 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
