siRNA Details
Gene Symbol: GRK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005308.2 | NP_005299.1 | 3 | 432 | ||
XM_005269707.1 | XP_005269764.1 | 3 | 429 | |||
XM_005269708.1 | XP_005269765.1 | 2 | 322 | |||
GenBank mRNA | AK298261.1 | - | 3 | 375 | ||
AK301450.1 | - | 2 | 123 | |||
AK309761.1 | - | 3 | 414 | |||
AK313012.1 | - | 3 | 279 | |||
BC064506.1 | - | 3 | 426 | 6482467 | ||
L15388.1 | - | 3 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
