siRNA Details
Gene Symbol: Ephb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159571.1 | NP_001153043.1 | 3 | 821 | ||
NM_010144.6 | NP_034274.4 | 3 | 821 | |||
GenBank mRNA | AK046884.1 | - | 3 | 759 | ||
AK049340.1 | - | 3 | 822 | |||
AK132392.1 | - | 3 | 821 | |||
AK143422.1 | - | 790 | ||||
BC013342.1 | - | 3 | 481 | |||
BC016505.1 | - | 3 | 773 | 3709926 | ||
BC090839.1 | - | 3 | 598 | |||
U06834.1 | - | 4 | 434 |
