siRNA Details
Gene Symbol: Evi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033711.1 | NP_001028883.1 | 2 | 1203 | ||
NM_010161.3 | NP_034291.1 | 2 | 390 | |||
GenBank mRNA | AK044098.1 | - | 2 | 266 | ||
AK046407.1 | - | 2 | 341 | |||
AK160302.1 | - | 2 | 307 | |||
AK162958.1 | - | 266 | ||||
AK170527.1 | - | 2 | 362 | |||
AK171332.1 | - | 1 | 2747 | |||
AK171594.1 | - | 2 | 308 | |||
BC038124.1 | - | 2 | 347 | 1396641 | ||
M34896.1 | - | 2 | 320 | |||
M34897.1 | - | 1 | 716 |
Gene Symbol: Gm21975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146023.4 | NP_666135.1 | 2 | 390 | ||
GenBank mRNA | BB626486.1 | - | 2 | 341 | ||
BY214628.1 | - | 2 | 230 | |||
BY221230.1 | - | 2 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
