siRNA Details
Gene Symbol: Ackr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010045.2 | NP_034175.2 | 2 | 1070 | ||
GenBank mRNA | AF016584.1 | - | 2 | 1011 | ||
AF109159.1 | - | 2 | 1070 | |||
AY289754.1 | - | 2 | 977 | |||
AY291295.1 | - | 2 | 977 | |||
BC005583.1 | - | 2 | 1069 | 3968984 |
