siRNA Details
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052854.3 | NP_443086.1 | 2 | 626 | ||
XM_006718380.3 | XP_006718443.1 | 2 | 603 | |||
GenBank mRNA | AB063321.1 | - | 2 | 247 | ||
AK314072.1 | - | 2 | 604 | |||
BC014097.1 | - | 2 | 269 | 4546950 | ||
BC015781.1 | - | 2 | 255 | 4856946 | ||
BQ953032.1 | - | 2 | 586 | |||
CF131922.1 | - | 2 | 622 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011957, AB017614, BC016447, AK142368.
Gene Ontology Categories:
